missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromonaphthalene, 99%

Catalog No. AAL1613403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1613403 1 g
AAL1613406 5 g
AAL1613414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1613403 Supplier Thermo Scientific Chemicals Supplier No. L1613403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13720-06-4 | C10H6Br2 | 285.97 g/mol

2,6-Dibromonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It reacts with hexylmagnesium bromide to form 2,6-Dihexyl-naphthalene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It reacts with hexylmagnesium bromide to form 2,6-Dihexyl-naphthalene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13720-06-4
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD01026462
InChI Key PJZDEYKZSZWFPX-UHFFFAOYSA-N
Synonym naphthalene, 2,6-dibromo, 2,6-dibromo-naphthalene, 2,6-dibromo naphthalene, 2.6-dibromonaphthalene, 2,6-?dibromonaphthalene, acmc-1c7c1, ksc490o3h, 2,6-dibromonaphthalene
PubChem CID 640591
IUPAC Name 2,6-dibromonaphthalene
SMILES BrC1=CC2=CC=C(Br)C=C2C=C1

Specifications

Melting Point 162°C to 167°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 285.97
Percent Purity 99%
Chemical Name or Material 2,6-Dibromonaphthalene
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only