missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide 98.0+%, TCI America™
$398.90 - $1189.27
Chemical Identifiers
| CAS | 1088205-02-0 |
|---|---|
| Molecular Formula | C30H36Br2N2O4 |
| Molecular Weight (g/mol) | 648.44 |
| MDL Number | MFCD27923032 |
| InChI Key | IMWDVKCSASRBSW-UHFFFAOYNA-N |
| PubChem CID | 71721476 |
| IUPAC Name | 2,9-dibromo-6,13-bis(2-ethylhexyl)-6,13-diazatetracyclo[6.6.2.0â´,¹â¶.0¹¹,¹âµ]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone |
| SMILES | CCCCC(CC)CN1C(=O)C2=CC(Br)=C3C(=O)N(CC(CC)CCCC)C(=O)C4=CC(Br)=C(C1=O)C2=C34 |
Chemical Identifiers
| 1088205-02-0 | |
| 648.44 | |
| IMWDVKCSASRBSW-UHFFFAOYNA-N | |
| 2,9-dibromo-6,13-bis(2-ethylhexyl)-6,13-diazatetracyclo[6.6.2.0â´,¹â¶.0¹¹,¹âµ]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone |
| C30H36Br2N2O4 | |
| MFCD27923032 | |
| 71721476 | |
| CCCCC(CC)CN1C(=O)C2=CC(Br)=C3C(=O)N(CC(CC)CCCC)C(=O)C4=CC(Br)=C(C1=O)C2=C34 |
Specifications
| 1088205-02-0 | |
| C30H36Br2N2O4 | |
| 1 g | |
| CCCCC(CC)CN1C(=O)C2=CC(Br)=C3C(=O)N(CC(CC)CCCC)C(=O)C4=CC(Br)=C(C1=O)C2=C34 | |
| 648.44 | |
| 648.44 | |
| Crystalline Powder |
| 245°C | |
| MFCD27923032 | |
| IMWDVKCSASRBSW-UHFFFAOYNA-N | |
| 2,9-dibromo-6,13-bis(2-ethylhexyl)-6,13-diazatetracyclo[6.6.2.0â´,¹â¶.0¹¹,¹âµ]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone | |
| 71721476 | |
| ≥98.0% (HPLC,N) | |
| 2,6-Dibromo-N,N′-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide |
Safety and Handling
TSCA : No