missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-N-chloro-p-benzoquinoneimine, 97%

Catalog No. p-4258930
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406470250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC406470250 Supplier Thermo Scientific Chemicals Supplier No. 406470250

CAS: 537-45-1 | C6H2Br2ClNO | 299.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 537-45-1
Molecular Formula C6H2Br2ClNO
Molecular Weight (g/mol) 299.34
InChI Key JYWKEVKEKOTYEX-UHFFFAOYSA-N
Synonym 2,6-dibromoquinone-4-chloroimide, 2,6-dibromo-4-chloroimino cyclohexa-2,5-dienone, bqc reagent, 2,6-dibromoquinone-4-chlorimide, 2,6-dibromoquinone chlorimide, 2,6-dibromoquinone chloroimide, 2,6-dibromoquinone chloroimine, n-chloro-2,6-dibromoquinoneimine, 2,5-cyclohexadien-1-one, 2,6-dibromo-4-chloroimino, 2,6-dibromo-p-benzoquinone-4-chlorimine
PubChem CID 10835
IUPAC Name 2,6-dibromo-4-chloroiminocyclohexa-2,5-dien-1-one
SMILES C1=C(C(=O)C(=CC1=NCl)Br)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 76.0°C to 83.0°C
Color Yellow to Ochre
Density 1.8100g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Iodometry)
Packaging Glass bottle
Quantity 25 g
Merck Index 15,3031
Specific Gravity 1.81
Formula Weight 299.34
Percent Purity 97%
Physical Form Powder or Fine Crystalline Needles
Chemical Name or Material 2, 6-Dibromo-N-chloro-p-benzoquinoneimine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Heating may cause a fire.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 208-667-1
Recommended Storage Product may discolor to orange-brown on storage

RUO – Research Use Only