missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-nitrotoluene, 95%

Catalog No. p-7046090
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1310206 5 g
AAL1310214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1310206 Supplier Thermo Scientific Chemicals Supplier No. L1310206

missing translation for 'validMainframeMsg'

CAS: 110127-07-6 | C7H5Br2NO2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110127-07-6
Molecular Formula C7H5Br2NO2
MDL Number MFCD00151807

Specifications

Melting Point 54°C to 57°C
Quantity 5 g
Beilstein 3259701
Formula Weight 294.94
Percent Purity 95%
Chemical Name or Material 2,6-Dibromo-4-nitrotoluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only