missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromo-4,4-di-n-octyldithieno[3,2-b:2',3'-d]silole 98.0+%, TCI America™
Supplier: TCI America D4799100MG
Specifications
| 2,6-Dibromo-4,4-di-n-octyldithieno[3,2-b:2′,3′-d]silole | |
| C24H36Br2S2Si | |
| 100 mg | |
| PVZQCRDPHHCCRP-UHFFFAOYSA-N | |
| 576.565 | |
| 576.57 | |
| Crystalline Powder |
| 1160106-14-8 | |
| MFCD26793508 | |
| 2,6-Dibromo-4,4-di-n-octyl-4H-silolo[3,2-b:4,5-b′C]dithiophene | |
| CCCCCCCC[Si]1(C2=C(C3=C1C=C(S3)Br)SC(=C2)Br)CCCCCCCC | |
| 68012321 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1160106-14-8 | |
| 576.565 | |
| PVZQCRDPHHCCRP-UHFFFAOYSA-N | |
| 68012321 |
| C24H36Br2S2Si | |
| MFCD26793508 | |
| 2,6-Dibromo-4,4-di-n-octyl-4H-silolo[3,2-b:4,5-b′C]dithiophene | |
| CCCCCCCC[Si]1(C2=C(C3=C1C=C(S3)Br)SC(=C2)Br)CCCCCCCC |
Safety and Handling
TSCA : No