missing translation for 'onlineSavingsMsg'
Learn More

2,6-dibromo-3-chloro-4-fluoroaniline, Thermo Scientific™

Catalog No. BTB04528EB
Change view
Click to view available options
Quantity:
25g
50g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB04528EB 25g Amber Glass Bottle
BTB04528EE 50g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB04528EB Supplier Thermo Scientific Chemicals Supplier No. BTB04528EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175135-09-8
Molecular Formula C6H3Br2ClFN
Molecular Weight (g/mol) 303.353
InChI Key PRZBEZGEEPNKRO-UHFFFAOYSA-N
Synonym 3-chloro-2,6-dibromo-4-fluoroaniline, 2,6-dibromo-3-chloro-4-fluorobenzenamine, 1-chloro-2,6-dibromo-4-fluoroaniline, benzenamine, 2,6-dibromo-3-chloro-4-fluoro, benzenamine,2,6-dibromo-3-chloro-4-fluoro, pubchem2948, acmc-1c6hf, 2,6-dibromo-3-chloro-4-fluorophenylamine
PubChem CID 2736480
IUPAC Name 2,6-dibromo-3-chloro-4-fluoroaniline
SMILES C1=C(C(=C(C(=C1Br)N)Br)Cl)F

Specifications

Packaging Amber Glass Bottle
Quantity 25g
Formula Weight 303.36
Percent Purity 97%
Chemical Name or Material 2, 6-dibromo-3-chloro-4-fluoroaniline
Hazard Category GHS Signal Word: Warning