missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopurine, 98%

Catalog No. p-7022746
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1564903 1 g
AAA1564906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1564903 Supplier Thermo Scientific Chemicals Supplier No. A1564903
Only null left

CAS: 1904-98-9 | C5H6N6 | 150.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1904-98-9
Molecular Formula C5H6N6
Molecular Weight (g/mol) 150.145
MDL Number MFCD00213668
InChI Key MSSXOMSJDRHRMC-UHFFFAOYSA-N
Synonym 2,6-diaminopurine, 9h-purine-2,6-diamine, 2-aminoadenine, 1h-purine-2,6-diamine, 2,6-diamino-9h-purine, purine, 2,6-diamino, purine-2,6-diyldiamine, unii-49p95bau4z, ccris 923, purine-2,6-diamine
PubChem CID 30976
ChEBI CHEBI:40235
IUPAC Name 7H-purine-2,6-diamine
SMILES C1=NC2=C(N1)C(=NC(=N2)N)N

Specifications

Melting Point ∼302°C
Quantity 1 g
Beilstein 9624
Merck Index 14,2985
Formula Weight 150.14
Percent Purity 98%
Chemical Name or Material 2,6-Diaminopurine
Hazard Category H320-H351
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P264b-P280-P305+P351+P338-P308+P313-P501c
EINECSNumber 217-605-2
RTECSNumber UO7524000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only