Learn More
2,6-Diacetylpyridine, 99%
CAS: 1129-30-2 | C9H9NO2 | 163.18 g/mol
Supplier: Thermo Scientific Chemicals B2567806
Description
2,6-Diacetylpyridine is used as intermediate in organic synthesis. It is a precursor to ligands in coordination chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Diacetylpyridine is used as intermediate in organic synthesis. It is a precursor to ligands in coordination chemistry.
Solubility
Soluble in water, chloroform, and DMSO.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
| 2,6-Diacetylpyridine | |
| 78°C to 82°C | |
| MFCD00006304 | |
| 2,6-diacetylpyridine, 1,1'-pyridine-2,6-diyl diethanone, 1-6-acetylpyridin-2-yl ethanone, pyridine-2,6-diacetyl, 2,6-diacetyl pyridine, 1-6-acetyl-pyridin-2-yl-ethanone, ethanone, 1,1'-2,6-pyridinediyl bis, 1-6-acetylpyridin-2-yl ethan-1-one, 2,6diacetylpyridine, pubchem3144 | |
| BEZVGIHGZPLGBL-UHFFFAOYSA-N | |
| 1-(6-acetylpyridin-2-yl)ethanone | |
| 70790 | |
| 99% |
| 1129-30-2 | |
| C9H9NO2 | |
| 5 g | |
| Soluble in water,chloroform,and DMSO. | |
| CC(=O)C1=CC=CC(=N1)C(C)=O | |
| 163.18 | |
| 163.18 |
Chemical Identifiers
| 1129-30-2 | |
| 163.18 | |
| BEZVGIHGZPLGBL-UHFFFAOYSA-N | |
| 70790 | |
| CC(=O)C1=CC=CC(=N1)C(C)=O |
| C9H9NO2 | |
| MFCD00006304 | |
| 2,6-diacetylpyridine, 1,1'-pyridine-2,6-diyl diethanone, 1-6-acetylpyridin-2-yl ethanone, pyridine-2,6-diacetyl, 2,6-diacetyl pyridine, 1-6-acetyl-pyridin-2-yl-ethanone, ethanone, 1,1'-2,6-pyridinediyl bis, 1-6-acetylpyridin-2-yl ethan-1-one, 2,6diacetylpyridine, pubchem3144 | |
| 1-(6-acetylpyridin-2-yl)ethanone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 214-442-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only