missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diacetylpyridine, 99%

Catalog No. AC330860250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330860250 25 g Glass bottle
AC330860010 1 g Glass bottle
AC330860050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC330860250 Supplier Thermo Scientific Chemicals Supplier No. 330860250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1129-30-2 | C9H9NO2 | 163.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1129-30-2
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00006304
InChI Key BEZVGIHGZPLGBL-UHFFFAOYSA-N
Synonym 2,6-diacetylpyridine, 1,1'-pyridine-2,6-diyl diethanone, 1-6-acetylpyridin-2-yl ethanone, pyridine-2,6-diacetyl, 2,6-diacetyl pyridine, 1-6-acetyl-pyridin-2-yl-ethanone, ethanone, 1,1'-2,6-pyridinediyl bis, 1-6-acetylpyridin-2-yl ethan-1-one, 2,6diacetylpyridine, pubchem3144
PubChem CID 70790
IUPAC Name 1-(6-acetylpyridin-2-yl)ethanone
SMILES CC(=O)C1=CC=CC(=N1)C(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 78°C to 82°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H3N)(COCH3)2
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform,dimethylsulfoxide
Formula Weight 163.18
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 2, 6-Diacetylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-442-9

RUO – Research Use Only