missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diacetylpyridine, 99%

Catalog No. AAB2567814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2567814 25 g
AAB2567803 1 g
AAB2567806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2567814 Supplier Thermo Scientific Chemicals Supplier No. B2567814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1129-30-2 | C9H9NO2 | 163.18 g/mol

2,6-Diacetylpyridine is used as intermediate in organic synthesis. It is a precursor to ligands in coordination chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diacetylpyridine is used as intermediate in organic synthesis. It is a precursor to ligands in coordination chemistry.

Solubility
Soluble in water, chloroform, and DMSO.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 1129-30-2
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00006304
InChI Key BEZVGIHGZPLGBL-UHFFFAOYSA-N
Synonym 2,6-diacetylpyridine, 1,1'-pyridine-2,6-diyl diethanone, 1-6-acetylpyridin-2-yl ethanone, pyridine-2,6-diacetyl, 2,6-diacetyl pyridine, 1-6-acetyl-pyridin-2-yl-ethanone, ethanone, 1,1'-2,6-pyridinediyl bis, 1-6-acetylpyridin-2-yl ethan-1-one, 2,6diacetylpyridine, pubchem3144
PubChem CID 70790
IUPAC Name 1-(6-acetylpyridin-2-yl)ethanone
SMILES CC(=O)C1=CC=CC(=N1)C(C)=O

Specifications

Melting Point 78°C to 82°C
Quantity 25 g
Solubility Information Soluble in water,chloroform,and DMSO.
Formula Weight 163.18
Percent Purity 99%
Chemical Name or Material 2,6-Diacetylpyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-442-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only