missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-p-benzoquinone, 98%

CAS: 719-22-2 | C14H20O2 | 220.31 g/mol

$155.47 - $523.69

Chemical Identifiers

CAS 719-22-2
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.31
InChI Key RDQSIADLBQFVMY-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-p-benzoquinone, 2,6-di-tert-butyl-1,4-benzoquinone, 2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,6-di-tert-butylbenzoquinone, 2,6-di-tert-butylquinone, 2,6-di-t-butyl-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-bis 1,1-dimethylethyl, p-benzoquinone, 2,6-di-tert-butyl, unii-4c9d8l0y0t, 2,6-di tert-butyl benzo-1,4-quinone
PubChem CID 12867
IUPAC Name 2,6-ditert-butylcyclohexa-2,5-diene-1,4-dione
SMILES CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC167340050
View Documents Promotion Details
N/A 5 g
Each for $155.47
Only null left
 
AC167340250
View Documents Promotion Details
N/A 25 g
Each for $523.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

719-22-2
220.31
2,6-di-tert-butyl-p-benzoquinone, 2,6-di-tert-butyl-1,4-benzoquinone, 2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,6-di-tert-butylbenzoquinone, 2,6-di-tert-butylquinone, 2,6-di-t-butyl-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-bis 1,1-dimethylethyl, p-benzoquinone, 2,6-di-tert-butyl, unii-4c9d8l0y0t, 2,6-di tert-butyl benzo-1,4-quinone
2,6-ditert-butylcyclohexa-2,5-diene-1,4-dione
C14H20O2
RDQSIADLBQFVMY-UHFFFAOYSA-N
12867
CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C

Specifications

719-22-2
75.0°C to 80.0°C (0.7 mmHg)
97.5% min. (GC)
[(CH3)3C]2C6H2(=O)2
2,6-di-tert-butyl-p-benzoquinone, 2,6-di-tert-butyl-1,4-benzoquinone, 2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,6-di-tert-butylbenzoquinone, 2,6-di-tert-butylquinone, 2,6-di-t-butyl-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-bis 1,1-dimethylethyl, p-benzoquinone, 2,6-di-tert-butyl, unii-4c9d8l0y0t, 2,6-di tert-butyl benzo-1,4-quinone
CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C
220.31
220.31
2,6-Di-tert-butyl-p-benzoquinone
65.0°C to 68.0°C
Authentic
C14H20O2
09,139
RDQSIADLBQFVMY-UHFFFAOYSA-N
2,6-ditert-butylcyclohexa-2,5-diene-1,4-dione
12867
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 211-946-

SDS
Documents

Documents

RUO – Research Use Only