missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-p-benzoquinone, 98+%

Catalog No. AAA1309106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1309106 5 g
AAA1309114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1309106 Supplier Thermo Scientific Chemicals Supplier No. A1309106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 719-22-2 | C14H20O2 | 220.312 g/mol

2,6-Di-tert-butyl-1,4-benzoquinone was used as an antioxidant to study the elimination rate of micro pollutant from storm and waste water. It was also used as a metabolite of butylated hydroxytoluene. It is used as a treatment agent to improve germination and plant health.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Di-tert-butyl-1,4-benzoquinone was used as an antioxidant to study the elimination rate of micro pollutant from storm and waste water. It was also used as a metabolite of butylated hydroxytoluene. It is used as a treatment agent to improve germination and plant health.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 719-22-2
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00001601
InChI Key RDQSIADLBQFVMY-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-p-benzoquinone, 2,6-di-tert-butyl-1,4-benzoquinone, 2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,6-di-tert-butylbenzoquinone, 2,6-di-tert-butylquinone, 2,6-di-t-butyl-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-bis 1,1-dimethylethyl, p-benzoquinone, 2,6-di-tert-butyl, unii-4c9d8l0y0t, 2,6-di tert-butyl benzo-1,4-quinone
PubChem CID 12867
IUPAC Name 2,6-ditert-butylcyclohexa-2,5-diene-1,4-dione
SMILES CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C

Specifications

Melting Point 65°C to 69°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1911512
Solubility Information Insoluble in water.
Formula Weight 220.31
Percent Purity ≥98%
Chemical Name or Material 2,6-Di-tert-butyl-p-benzoquinone
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 211-946-0
RTECSNumber DK3970000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only