missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylpyridine, 98%

CAS: 38222-83-2 | C14H23N | 205.35 g/mol

$268.35 - $268.35

Chemical Identifiers

CAS 38222-83-2
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.35
MDL Number MFCD00006305
InChI Key HVHZEKKZMFRULH-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methylpyridine, dtbmp, pyridine, 2,6-bis 1,1-dimethylethyl-4-methyl, 2,6-di-t-butyl-4-methylpyridine, 2,6-ditert-butyl-4-methyl-pyridine, 2,6-bis tert-butyl-4-methylpyridine, pubchem10244, acmc-209iyr, ksc490s2n, 2,6-di-t-butyl 4-methylpyridine
PubChem CID 98898
IUPAC Name 2,6-ditert-butyl-4-methylpyridine
SMILES CC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC224520050
SDS View Documents
Thermo Scientific Chemicals
224520050
5 g Glass bottle
Each for $268.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38222-83-2
205.35
HVHZEKKZMFRULH-UHFFFAOYSA-N
98898
CC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C
C14H23N
MFCD00006305
2,6-di-tert-butyl-4-methylpyridine, dtbmp, pyridine, 2,6-bis 1,1-dimethylethyl-4-methyl, 2,6-di-t-butyl-4-methylpyridine, 2,6-ditert-butyl-4-methyl-pyridine, 2,6-bis tert-butyl-4-methylpyridine, pubchem10244, acmc-209iyr, ksc490s2n, 2,6-di-t-butyl 4-methylpyridine
2,6-ditert-butyl-4-methylpyridine

Specifications

38222-83-2
100.0
Colorless to Yellow
83°C
97.5% min. (GC)
C14H23N
[(CH3)3C]2C5H2NCH3
5 g
09,141; 10,123; 11,160; 16,110
2,6-di-tert-butyl-4-methylpyridine, dtbmp, pyridine, 2,6-bis 1,1-dimethylethyl-4-methyl, 2,6-di-t-butyl-4-methylpyridine, 2,6-ditert-butyl-4-methyl-pyridine, 2,6-bis tert-butyl-4-methylpyridine, pubchem10244, acmc-209iyr, ksc490s2n, 2,6-di-t-butyl 4-methylpyridine
CC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C
205.35
205.34
Liquid or Low Melting Solid
97.5
33°C to 36°C
233°C
Authentic
Glass bottle
1.4760 to 1.478
MFCD00006305
20, V,6, 190
15, 3042
HVHZEKKZMFRULH-UHFFFAOYSA-N
2,6-ditert-butyl-4-methylpyridine
98898
98%
2, 6-Di-tert-butyl-4-methylpyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a P

GHS Signal Word: Warning

EINECSNumber : 253-834-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only