missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylpyridine, 98%

Catalog No. AC224520250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC224520250 25 g Glass bottle
AC224520050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC224520250 Supplier Thermo Scientific Chemicals Supplier No. 224520250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38222-83-2 | C14H23N | 205.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 38222-83-2
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.35
MDL Number MFCD00006305
InChI Key HVHZEKKZMFRULH-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methylpyridine, dtbmp, pyridine, 2,6-bis 1,1-dimethylethyl-4-methyl, 2,6-di-t-butyl-4-methylpyridine, 2,6-ditert-butyl-4-methyl-pyridine, 2,6-bis tert-butyl-4-methylpyridine, pubchem10244, acmc-209iyr, ksc490s2n, 2,6-di-t-butyl 4-methylpyridine
PubChem CID 98898
IUPAC Name 2,6-ditert-butyl-4-methylpyridine
SMILES CC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 36°C
Color Colorless to Yellow
Boiling Point 233°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3C]2C5H2NCH3
Refractive Index 1.4760 to 1.478
Quantity 25 g
Beilstein 20, V,6, 190
Fieser 09,141; 10,123; 11,160; 16,110
Merck Index 15, 3042
Formula Weight 205.34
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2, 6-Di-tert-butyl-4-methylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a P
EINECSNumber 253-834-4

RUO – Research Use Only