missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2,6-Di-tert-butyl-4-methylphenol

Catalog No. B13781KG
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13781KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B13781KG Supplier Sigma Aldrich Supplier No. B13781KG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,6-Di-tert-butyl-4-methylphenol

Specifications

CAS 128-37-0
Melting Point 69°C to 73°C (lit.)
Boiling Point 265°C (lit.)
Molecular Formula C15H24O
Linear Formula [(CH3)3 C]2C6H2(CH3)OH
MDL Number MFCD00011644
Quantity 1 kg
Synonym 2,6-Di-tert-butyl-p-cresol; BHT; Butylated hydroxytoluene; Butylhydroxytoluene; DBPC
Molecular Weight (g/mol) 220.35
Percent Purity ≥99.0% (GC)