missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-1,4-benzoquinone 98.0+%, TCI America™

Catalog No. D22565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22565G 5 g
D225625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22565G Supplier TCI America Supplier No. D22565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 719-22-2
Molecular Formula C14H20O2
Molecular Weight (g/mol) 220.312
MDL Number MFCD00001601
InChI Key RDQSIADLBQFVMY-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-p-benzoquinone, 2,6-di-tert-butyl-1,4-benzoquinone, 2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,6-di-tert-butylbenzoquinone, 2,6-di-tert-butylquinone, 2,6-di-t-butyl-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-bis 1,1-dimethylethyl, p-benzoquinone, 2,6-di-tert-butyl, unii-4c9d8l0y0t, 2,6-di tert-butyl benzo-1,4-quinone
PubChem CID 12867
IUPAC Name 2,6-ditert-butylcyclohexa-2,5-diene-1,4-dione
SMILES CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C

Specifications

Melting Point 67°C
Quantity 5 g
Formula Weight 220.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di-tert-butyl-1,4-benzoquinone
RTECSNumber DK3970000
TSCA Yes