missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di-tert-butyl-1,4-benzoquinone 98.0+%, TCI America™
Supplier: TCI America D225625G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C14H20O2 | |
| MFCD00001601 | |
| 2,6-di-tert-butyl-p-benzoquinone, 2,6-di-tert-butyl-1,4-benzoquinone, 2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,6-di-tert-butylbenzoquinone, 2,6-di-tert-butylquinone, 2,6-di-t-butyl-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-bis 1,1-dimethylethyl, p-benzoquinone, 2,6-di-tert-butyl, unii-4c9d8l0y0t, 2,6-di tert-butyl benzo-1,4-quinone | |
| 2,6-ditert-butylcyclohexa-2,5-diene-1,4-dione |
Specifications
| 2,6-Di-tert-butyl-1,4-benzoquinone | |
| 67°C | |
| 25 g | |
| 2,6-di-tert-butyl-p-benzoquinone, 2,6-di-tert-butyl-1,4-benzoquinone, 2,6-di-tert-butylcyclohexa-2,5-diene-1,4-dione, 2,6-di-tert-butylbenzoquinone, 2,6-di-tert-butylquinone, 2,6-di-t-butyl-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-bis 1,1-dimethylethyl, p-benzoquinone, 2,6-di-tert-butyl, unii-4c9d8l0y0t, 2,6-di tert-butyl benzo-1,4-quinone | |
| CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C | |
| 220.312 | |
| 220.31 | |
| Crystalline Powder |
| 719-22-2 | |
| C14H20O2 | |
| MFCD00001601 | |
| RDQSIADLBQFVMY-UHFFFAOYSA-N | |
| 2,6-ditert-butylcyclohexa-2,5-diene-1,4-dione | |
| 12867 | |
| ≥98.0% (GC) |
Safety and Handling
RTECSNumber : DK3970000
TSCA : Yes