missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di(2H-1,2,3-triazol-4-yl)pyridine 98.0+%, TCI America™

Catalog No. D4879200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4879200MG 200 mg
D48791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4879200MG Supplier TCI America Supplier No. D4879200MG
Only null left

Chemical Identifiers

CAS 1542234-40-1
Molecular Formula C9H7N7
Molecular Weight (g/mol) 213.204
InChI Key GTUXTIISPOXPEF-UHFFFAOYSA-N
PubChem CID 90693344
IUPAC Name 2,6-bis(2H-triazol-4-yl)pyridine
SMILES C1=CC(=NC(=C1)C2=NNN=C2)C3=NNN=C3

Specifications

Melting Point 285°C
Color White-Yellow
Quantity 200 mg
Formula Weight 213.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di(2H-1,2,3-triazol-4-yl)pyridine