missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(p-tolyl)pyridine, 97%

CAS: 14435-88-2 | C19H17N | 259.352 g/mol

$59.30 - $197.71

Chemical Identifiers

CAS 14435-88-2
Molecular Formula C19H17N
Molecular Weight (g/mol) 259.352
MDL Number MFCD00006282
InChI Key BLRYHCTZTSRBNB-UHFFFAOYSA-N
PubChem CID 84435
IUPAC Name 2,6-bis(4-methylphenyl)pyridine
SMILES CC1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=C(C=C3)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2489403
View Documents
Thermo Scientific Chemicals
B2489403
1 g
Each for $59.30
Only null left
 
AAB2489406
View Documents
Thermo Scientific Chemicals
B2489406
5 g
Each for $197.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14435-88-2
259.352
BLRYHCTZTSRBNB-UHFFFAOYSA-N
2,6-bis(4-methylphenyl)pyridine
C19H17N
MFCD00006282
84435
CC1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=C(C=C3)C

Specifications

14435-88-2
C19H17N
1 g
BLRYHCTZTSRBNB-UHFFFAOYSA-N
2,6-bis(4-methylphenyl)pyridine
84435
97%
164°C to 166°C
MFCD00006282
2,6-bis p-tolyl pyridine, 2,6-di 4-methylphenyl pyridine, 2,6-di-p-tolylpyridine, 2,6-bis 4-methylphenyl pyridine, 2,6 di-p-tolylpyridine, 2,6-di-p-tolyl-pyridine, pyridine,2,6-di-p-tolyl, pyridine, 2,6-di-p-tolyl, 2,6-di-4-methylphenyl pyridine, pyridine, 2,6-bis 4-methylphenyl
CC1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=C(C=C3)C
259.352
259.35
2,6-Bis(p-tolyl)pyridine
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 238-409-3

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only