missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(hydroxymethyl)naphthalene 98.0+%, TCI America™

Catalog No. p-7122925
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33991G 1 g
B33995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33991G Supplier TCI America Supplier No. B33991G

Chemical Identifiers

CAS 5859-93-8
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
InChI Key SSFGHKDDKYEERH-UHFFFAOYSA-N
PubChem CID 11788722
IUPAC Name [6-(hydroxymethyl)naphthalen-2-yl]methanol
SMILES C1=CC2=C(C=CC(=C2)CO)C=C1CO

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 1 g
Formula Weight 188.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(hydroxymethyl)naphthalene