missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(diphenylamino)anthraquinone 96.0+%, TCI America™

Catalog No. B5092200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5092200MG 200 mg
B50921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5092200MG Supplier TCI America Supplier No. B5092200MG
Only null left

Chemical Identifiers

CAS 868850-50-4
Molecular Formula C38H26N2O2
Molecular Weight (g/mol) 542.64
MDL Number MFCD20040460
InChI Key TZENEWLXCXPNFX-UHFFFAOYSA-N
PubChem CID 58927077
IUPAC Name 2,6-bis(diphenylamino)-9,10-dihydroanthracene-9,10-dione
SMILES O=C1C2=C(C=C(C=C2)N(C2=CC=CC=C2)C2=CC=CC=C2)C(=O)C2=C1C=C(C=C2)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 200 mg
Formula Weight 542.64
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(diphenylamino)anthraquinone