missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(chloromethyl)pyridine 98.0+%, TCI America™

Catalog No. B14495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14495G 5 g
B144925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14495G Supplier TCI America Supplier No. B14495G

Chemical Identifiers

CAS 3099-28-3
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00142844
InChI Key IWQNFYRJSVJWQA-UHFFFAOYSA-N
PubChem CID 316654
IUPAC Name 2,6-bis(chloromethyl)pyridine
SMILES C1=CC(=NC(=C1)CCl)CCl

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 176.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(chloromethyl)pyridine