missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b']dithiophene 97.0+%, TCI America™
$445.51 - $1532.25
Chemical Identifiers
| CAS | 861398-06-3 |
|---|---|
| Molecular Formula | C22H28B2O4S2 |
| Molecular Weight (g/mol) | 442.202 |
| InChI Key | NTSDQKBGUDHDGQ-UHFFFAOYSA-N |
| PubChem CID | 70976318 |
| IUPAC Name | 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thieno[2,3-f][1]benzothiol-6-yl]-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC4=C(C=C(S4)B5OC(C(O5)(C)C)(C)C)C=C3S2 |
Chemical Identifiers
| 861398-06-3 | |
| 442.202 | |
| 70976318 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC4=C(C=C(S4)B5OC(C(O5)(C)C)(C)C)C=C3S2 |
| C22H28B2O4S2 | |
| NTSDQKBGUDHDGQ-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thieno[2,3-f][1]benzothiol-6-yl]-1,3,2-dioxaborolane |
Specifications
| 861398-06-3 | |
| 1 g | |
| NTSDQKBGUDHDGQ-UHFFFAOYSA-N | |
| 4,4,5,5-tetramethyl-2-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thieno[2,3-f][1]benzothiol-6-yl]-1,3,2-dioxaborolane | |
| 70976318 | |
| ≥97.0% (GC) | |
| 2,6-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[1,2-b:4,5-b′]dithiophene |
| C22H28B2O4S2 | |
| Benzo[1,2-b:4,5-b′C]dithiophene-2,6-diboronic Acid Bis(pinacol) Ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC4=C(C=C(S4)B5OC(C(O5)(C)C)(C)C)C=C3S2 | |
| 442.202 | |
| 442.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No