missing translation for 'onlineSavingsMsg'
Learn More

2,6,8-Trimethyl-4-nonanol (threo- and erythro- mixture), TCI America™

Catalog No. T22795G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22795G Supplier TCI America Supplier No. T22795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-17-1
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.339
MDL Number MFCD00026491
InChI Key LFEHSRSSAGQWNI-UHFFFAOYSA-N
Synonym 2,6,8-trimethyl-4-nonanol, 4-nonanol, 2,6,8-trimethyl, 4-hydroxy-2,6,8-trimethylnonane, 2,6,8-trimethylnonanol-4, 2,4,8-trimethyl-6-nonanol, dsstox_cid_9159, dsstox_rid_78690, dsstox_gsid_29159, acmc-1bphg, 2,8-trimethylnonanol-4
PubChem CID 61056
IUPAC Name 2,6,8-trimethylnonan-4-ol
SMILES CC(C)CC(C)CC(CC(C)C)O

Specifications

Color Yellow
Boiling Point 113°C
Quantity 5 g
Formula Weight 186.34
Physical Form Liquid
Chemical Name or Material 2,6,8-Trimethyl-4-nonanol (threo- and erythro- mixture)
EINECSNumber (2)-0217
RTECSNumber RA7950000
TSCA Yes