missing translation for 'onlineSavingsMsg'
Learn More

2,6,8-Trimethyl-4-nonanol, erythro + threo, 90+%, Thermo Scientific Chemicals

Catalog No. AAB2083006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2083006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2083006 Supplier Thermo Scientific Chemicals Supplier No. B2083006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 123-17-1 | Molecular Formula: C12H26O | Molecular Weight: 186.339 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-17-1
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.339
MDL Number MFCD00026491
InChI Key LFEHSRSSAGQWNI-UHFFFAOYSA-N
Synonym 2,6,8-trimethyl-4-nonanol, 4-nonanol, 2,6,8-trimethyl, 4-hydroxy-2,6,8-trimethylnonane, 2,6,8-trimethylnonanol-4, 2,4,8-trimethyl-6-nonanol, dsstox_cid_9159, dsstox_rid_78690, dsstox_gsid_29159, acmc-1bphg, 2,8-trimethylnonanol-4
PubChem CID 61056
IUPAC Name 2,6,8-trimethylnonan-4-ol
SMILES CC(C)CC(C)CC(CC(C)C)O

Specifications

Boiling Point 123°C (20 mmHg)
Refractive Index 1.435
Quantity 5 g
Formula Weight 184.32
Percent Purity ≥90%
Chemical Name or Material 2,6,8-Trimethyl-4-nonanol, erythro + threo
EINECSNumber 204-606-8
RTECSNumber RA7950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only