missing translation for 'onlineSavingsMsg'
Learn More

2,6,10,14-Tetramethylpentadecane 95.0+%, TCI America™

Catalog No. T01495ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01495ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T01495ML Supplier TCI America Supplier No. T01495ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1921-70-6
Molecular Formula C19H40
Molecular Weight (g/mol) 268.529
MDL Number MFCD00008952
InChI Key XOJVVFBFDXDTEG-UHFFFAOYSA-N
Synonym pristane, norphytane, bute hydrocarbon, norphytan, pristan, pentadecane, 2,6,10,14-tetramethyl, meso-pristane, 2,6,10,10-tetramethylpentadecane, meso-form, norphytane, robuoy
PubChem CID 15979
ChEBI CHEBI:53181
IUPAC Name 2,6,10,14-tetramethylpentadecane
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point -99°C
Color Yellow
Boiling Point 299°C
Quantity 5 mL
Formula Weight 268.53
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6,10,14-Tetramethylpentadecane
EINECSNumber (2)-0010
RTECSNumber RZ1880000
TSCA Yes