missing translation for 'onlineSavingsMsg'
Learn More

2,6,10,14-Tetramethylpentadecane, 95%

Catalog No. p-198861
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138460050 5 mL Glass bottle
AC138460500 50 mL Glass bottle
AC138462500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC138460050 Supplier Thermo Scientific Chemicals Supplier No. 138460050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1921-70-6 | C19H40 | 268.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1921-70-6
Molecular Formula C19H40
Molecular Weight (g/mol) 268.51
InChI Key XOJVVFBFDXDTEG-UHFFFAOYSA-N
Synonym pristane, norphytane, bute hydrocarbon, norphytan, pristan, pentadecane, 2,6,10,14-tetramethyl, meso-pristane, 2,6,10,10-tetramethylpentadecane, meso-form, norphytane, robuoy
PubChem CID 15979
ChEBI CHEBI:53181
IUPAC Name 2,6,10,14-tetramethylpentadecane
SMILES CC(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.7800g/mL
Boiling Point 296°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4369 to 1.4389
Linear Formula (CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)(CH2)3CH(CH3)2
Quantity 5 mL
Merck Index 15,787
Specific Gravity 0.78
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,petroleum ether,benzene,,chloroform and carbon tetrachloride
Viscosity 5 mPa.s (25°C)
Formula Weight 268.51
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2,6,10,14-Tetramethylpentadecane, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-650-8
RTECSNumber RZ1880000
TSCA TSCA

RUO – Research Use Only