missing translation for 'onlineSavingsMsg'
Learn More

2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine 98.0+%, TCI America™

Catalog No. p-7137821
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3638200MG 200 mg
T36381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3638200MG Supplier TCI America Supplier No. T3638200MG
Only null left

Chemical Identifiers

CAS 219508-27-7
Molecular Formula C8H5F3N4
Molecular Weight (g/mol) 214.151
MDL Number MFCD26403657
InChI Key MOGTVLAYAVGOJK-UHFFFAOYSA-N
Synonym 3-(Trifluoromethyl)-5-(2-pyridyl)-1,2,4-triazole
PubChem CID 20807129
IUPAC Name 2-[5-(trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine
SMILES C1=CC=NC(=C1)C2=NNC(=N2)C(F)(F)F

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 200 mg
Formula Weight 214.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[5-(Trifluoromethyl)-1H-1,2,4-triazol-3-yl]pyridine