missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium Tetrafluoroborate, TCI America™

Catalog No. N06345G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06345G Supplier TCI America Supplier No. N06345G

missing translation for 'validMainframeMsg'

[Coupling Reagent for Peptide]

Chemical Identifiers

CAS 125700-73-4
Molecular Formula C14H20BF4N3O3
Molecular Weight (g/mol) 365.14
MDL Number MFCD00167753
InChI Key DMTWWFCCBWWXSA-UHFFFAOYNA-N
Synonym 2-endo-5-norbornene-2,3-dicarboxymido-1,1,3,3-tetramethyluronium tetrafluoroborate, dimethylamino 1r,2s,6r,7s-3,5-dioxo-4-azatricyclo 5.2.1.0_,? dec-8-en-4-yl oxy methylidene dimethylazanium tetrafluoroborate
PubChem CID 91634054
IUPAC Name [(dimethylamino)({3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}oxy)methylidene]dimethylazanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN(C)C(ON1C(=O)C2C3CC(C=C3)C2C1=O)=[N+](C)C

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 365.14
Physical Form Crystalline Powder
Chemical Name or Material 2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium Tetrafluoroborate [Coupling Reagent fo