missing translation for 'onlineSavingsMsg'
Learn More

2,5-Lutidine 98.0+%, TCI America™

Catalog No. p-7131800
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
500 mL
Catalog No. Quantity
L00655ML 5 mL
L006525ML 25 mL
L0065500ML 500 mL
3 options

Catalog No. L00655ML

Supplier: TCI America L00655ML

Only null left

Chemical Identifiers

CAS 589-93-5
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006343
InChI Key XWKFPIODWVPXLX-UHFFFAOYSA-N
Synonym 2,5-Dimethylpyridine
PubChem CID 11526
IUPAC Name 2,5-dimethylpyridine
SMILES CC1=CC=C(C)N=C1

Specifications

Melting Point -6°C
Color Yellow
Boiling Point 157°C
Quantity 5 mL
UN Number 1993
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Lutidine
EINECSNumber (5)-0712
RTECSNumber OK9625000
TSCA Yes