Learn More
2,5-Hexanedione, 97%
Description
2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.
Solubility
Miscible with alcohol, water and diethyl ether.
Notes
Incompatible with strong oxidizing agents, strong bases, metals and strong reducing agents.
Spécifications
Spécifications
| Melting Point | -6°C |
| Density | 0.976 |
| Boiling Point | 192°C to 193°C |
| Flash Point | 78°C (172°F) |
| Refractive Index | 1.425 |
| Quantity | 25 g |
| Beilstein | 506525 |
| Merck Index | 14,71 |
| Solubility Information | Miscible with alcohol,water and diethyl ether. |
| Formula Weight | 114.15 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H227-H315-H319-H373 |
| Hazard Statement | GHS H Statement H361-H373-H302-H332-H315-H319-H335-H227 Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P260-P264b-P280-P302+P352-P305+P351+P338-P314-P332+P313-P362-P370+P378q-P501c |
| EINECSNumber | 203-738-3 |
| RTECSNumber | MO3150000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only