missing translation for 'onlineSavingsMsg'
Learn More

2,5-Furandicarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7129846
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07105G 5 g
F071025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07105G Supplier TCI America Supplier No. F07105G
Only null left

Chemical Identifiers

CAS 3238-40-2
Molecular Formula C6H4O5
Molecular Weight (g/mol) 156.09
MDL Number MFCD00016582
InChI Key CHTHALBTIRVDBM-UHFFFAOYSA-N
Synonym 2,5-furandicarboxylic acid, dehydromucic acid, furan-2,5-dicarbonsaeure, 2,5-furandicarboxylicacid, 2,5-furandicarboxylate, furane-alpha,alpha'-dicarboxylic acid, furan 2,5-dicarboxylic acid, furan-2,5-dicarboxylate, furane-.alpha.,.alpha.'-dicarboxylic acid, 2,5 furan dicarboxylic acid
PubChem CID 76720
ChEBI CHEBI:84212
IUPAC Name furan-2,5-dicarboxylic acid
SMILES OC(=O)C1=CC=C(O1)C(O)=O

Specifications

Melting Point 320°C
Color White-Yellow
Quantity 5 g
Formula Weight 156.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Furandicarboxylic Acid