missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Furandicarbonyl Dichloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 78°C |
| Color | White |
| Boiling Point | 245°C |
| Quantity | 5 g |
| UN Number | 3261 |
| Formula Weight | 192.98 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,5-Furandicarbonyl Dichloride |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |