missing translation for 'onlineSavingsMsg'
Learn More

2,5-Furandicarbonyl Dichloride 98.0+%, TCI America™

Catalog No. F10221G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10221G 1 g
F10225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10221G Supplier TCI America Supplier No. F10221G
Only null left

Chemical Identifiers

CAS 10375-34-5
Molecular Formula C6H2Cl2O3
Molecular Weight (g/mol) 192.979
MDL Number MFCD28044847
InChI Key PDSULNVJASBMLP-UHFFFAOYSA-N
PubChem CID 11480968
IUPAC Name furan-2,5-dicarbonyl chloride
SMILES C1=C(OC(=C1)C(=O)Cl)C(=O)Cl

Specifications

Melting Point 78°C
Color White
Boiling Point 245°C
Quantity 1 g
UN Number 3261
Formula Weight 192.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Furandicarbonyl Dichloride
TSCA No
Recommended Storage Refrigerator