missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diphenyloxazole 99.0+%, TCI America™

Catalog No. p-7126493
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D090225G 25 g
D0902100G 100 g
D0902500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D090225G Supplier TCI America Supplier No. D090225G
Only null left

Chemical Identifiers

CAS 92-71-7
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.26
MDL Number MFCD00005306
InChI Key CNRNYORZJGVOSY-UHFFFAOYSA-N
Synonym 2,5-diphenyloxazole, oxazole, 2,5-diphenyl, tritosol, 2,5-diphenyl-oxazole, ppo scintillator, dpo scintillator, usaf ek-6775, ppo van, ppo, ppo scintillator van
PubChem CID 7105
IUPAC Name 2,5-diphenyl-1,3-oxazole
SMILES O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 25 g
Formula Weight 221.26
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diphenyloxazole [for scintillation spectrometry]
EINECSNumber (5)-0192
RTECSNumber RP6825000
TSCA Yes