missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diphenyloxazole, 99%

Catalog No. AAA1065436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1065436 500 g
AAA1065414 25 g
AAA1065422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1065436 Supplier Thermo Scientific Chemicals Supplier No. A1065436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 92-71-7 | C15H11NO | 221.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92-71-7
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.26
MDL Number MFCD00005306
InChI Key CNRNYORZJGVOSY-UHFFFAOYSA-N
Synonym 2,5-diphenyloxazole, oxazole, 2,5-diphenyl, tritosol, 2,5-diphenyl-oxazole, ppo scintillator, dpo scintillator, usaf ek-6775, ppo van, ppo, ppo scintillator van
PubChem CID 7105
IUPAC Name 2,5-diphenyl-1,3-oxazole
SMILES O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 70°C to 74°C
Density 1.06
Boiling Point 360°C
Quantity 500 g
Beilstein 157021
Formula Weight 221.26
Percent Purity 99%
Chemical Name or Material 2,5-Diphenyloxazole
EINECSNumber 202-181-3
RTECSNumber RP6825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only