missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethylpyrrole 98.0+%, TCI America™
$74.56 - $210.63
Chemical Identifiers
| CAS | 625-84-3 |
|---|---|
| Molecular Formula | C6H9N |
| Molecular Weight (g/mol) | 95.145 |
| MDL Number | MFCD00005223 |
| InChI Key | PAPNRQCYSFBWDI-UHFFFAOYSA-N |
| Synonym | 2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole |
| PubChem CID | 12265 |
| IUPAC Name | 2,5-dimethyl-1H-pyrrole |
| SMILES | CC1=CC=C(N1)C |
Chemical Identifiers
| 625-84-3 | |
| 95.145 | |
| PAPNRQCYSFBWDI-UHFFFAOYSA-N | |
| 12265 | |
| CC1=CC=C(N1)C |
| C6H9N | |
| MFCD00005223 | |
| 2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole | |
| 2,5-dimethyl-1H-pyrrole |
Specifications
| 625-84-3 | |
| 169°C | |
| MFCD00005223 | |
| 2929 | |
| PAPNRQCYSFBWDI-UHFFFAOYSA-N | |
| 2,5-dimethyl-1H-pyrrole | |
| 12265 | |
| ≥98.0% (GC) | |
| 2,5-Dimethylpyrrole |
| Yellow | |
| C6H9N | |
| 5 mL | |
| 2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole | |
| CC1=CC=C(N1)C | |
| 95.145 | |
| 95.15 | |
| Liquid |
Safety and Handling
RTECSNumber : UX9465930
TSCA : No
Recommended Storage : Refrigerator