missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrrole, 97%

CAS: 625-84-3 | C6H9N | 95.14 g/mol

$90.46 - $90.46

Chemical Identifiers

CAS 625-84-3
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.14
MDL Number MFCD00005223
InChI Key PAPNRQCYSFBWDI-UHFFFAOYSA-N
Synonym 2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole
PubChem CID 12265
IUPAC Name 2,5-dimethyl-1H-pyrrole
SMILES CC1=CC=C(N1)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC116750050
SDS View Documents
Thermo Scientific Chemicals
116750050
5 g
Each for $90.46
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

625-84-3
95.14
PAPNRQCYSFBWDI-UHFFFAOYSA-N
12265
CC1=CC=C(N1)C
C6H9N
MFCD00005223
2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole
2,5-dimethyl-1H-pyrrole

Specifications

625-84-3
100.0
0.9350g/mL
54°C
96% min. (GC)
C6H9N
MFCD00005223
20, 172
2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole
PAPNRQCYSFBWDI-UHFFFAOYSA-N
2,5-dimethyl-1H-pyrrole
12265
97%
2, 5-Dimethylpyrrole
96.0
Orange-Yellow to Yellow
165.0°C (740.0 mmHg)
Authentic
Glass bottle
1.5040 to 1.5070
5 g
0.935
Solubility in water: insoluble
CC1=CC=C(N1)C
95.14
95.14
Crystalline Powder and/or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

GHS Signal Word: Danger

EINECSNumber : 210-913-8

Recommended Storage : Product may darken during storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only