missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylindole, 97%

Catalog No. AAH6687406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6687406 5 g
AAH6687403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6687406 Supplier Thermo Scientific Chemicals Supplier No. H6687406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1196-79-8 | C10H11N | 145.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1196-79-8
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00005621
InChI Key ZFLFWZRPMDXJCW-UHFFFAOYSA-N
Synonym 2,5-dimethylindole, 1h-indole, 2,5-dimethyl, indole, 2,5-dimethyl, unii-etu94v283m, 2,5-dimethyl indole, pubchem7330, acmc-1bvpm
PubChem CID 70965
IUPAC Name 2,5-dimethyl-1H-indole
SMILES CC1=CC2=C(C=C1)NC(=C2)C

Specifications

Melting Point 112°C to 113°C
Quantity 5 g
Beilstein 113222
Formula Weight 145.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Dimethylindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-816-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only