missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylhexane, 99%

Catalog No. AAA1522106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1522106 5 g
AAA1522114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1522106 Supplier Thermo Scientific Chemicals Supplier No. A1522106

CAS: 592-13-2 | C8H18 | 114.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 592-13-2
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00008951
InChI Key UWNADWZGEHDQAB-UHFFFAOYSA-N
Synonym biisobutyl, hexane, 2,5-dimethyl, diisobutyl, 2,5-dimethyl hexane, hexane,5-dimethyl, acmc-209mbd, 2,5-dimethylhexane, 2,5-dimethylhexane 5g, hexane, 2,5-dimethyl-8ci 9ci
PubChem CID 11592
IUPAC Name 2,5-dimethylhexane
SMILES CC(C)CCC(C)C

Specifications

Density 0.694
Boiling Point 107°C to 108°C
Flash Point 26°C (80°F)
Refractive Index 1.392
Quantity 5 g
UN Number UN3295
Beilstein 1696877
Formula Weight 114.23
Percent Purity 99%
Chemical Name or Material 2,5-Dimethylhexane
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H304-H226-H315-H336
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 209-745-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only