missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylfuran, 99%

Catalog No. p-117267
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC176030250 25 mL Glass bottle
AC176031000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC176030250 Supplier Thermo Scientific Chemicals Supplier No. 176030250
Only null left

CAS: 625-86-5 | C6H8O | 96.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 625-86-5
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00003250
InChI Key GSNUFIFRDBKVIE-UHFFFAOYSA-N
Synonym furan, 2,5-dimethyl, 2,5-dimethylfurane, 2,5-dimethyl-furan, 2,5-dimethyl furane, unii-dr5hl9oj7y, ccris 3158, 2,5-dimethyl furan, dr5hl9oj7y, 2.5-dimethyl furan, acmc-1avpg
PubChem CID 12266
IUPAC Name 2,5-dimethylfuran
SMILES CC1=CC=C(O1)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -62.0°C
Color Colorless to Amber
Density 0.9000g/mL
Boiling Point 92.0°C to 94.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Quantity 25 mL
Beilstein 17,41
Merck Index 15,3268
Specific Gravity 0.9
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in oil,organic solvents
Formula Weight 96.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 5-Dimethylfuran, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Highly flammable liquid and vapour.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 210-914-3
RTECSNumber LU0875000
TSCA TSCA
Recommended Storage May darken on storage

RUO – Research Use Only