missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzylamine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 93-48-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00039780
InChI Key LUJNPFWZXIGIPS-UHFFFAOYSA-N
Synonym 2,5-dimethylbenzylamine, 2,5-dimethylphenyl methanamine, 1-2,5-dimethylphenyl methanamine, benzenemethanamine, 2,5-dimethyl, 2,5-dimethyl-benzylamine, 2,5-dimethylbenzenemethanamine, 2,5-dimethylphenyl methylamine, 2,5-dimethylphenyl methylamine, 2-methylamino-p-xylene, 2, 5-dimethylbenzylamine, 2,5-dimethylbenzylam
PubChem CID 66735
IUPAC Name (2,5-dimethylphenyl)methanamine
SMILES CC1=CC(=C(C=C1)C)CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350860010
SDS View Documents
Thermo Scientific Chemicals
350860010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC350860050
SDS View Documents
Thermo Scientific Chemicals
350860050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

93-48-1
135.21
LUJNPFWZXIGIPS-UHFFFAOYSA-N
66735
CC1=CC(=C(C=C1)C)CN
C9H13N
MFCD00039780
2,5-dimethylbenzylamine, 2,5-dimethylphenyl methanamine, 1-2,5-dimethylphenyl methanamine, benzenemethanamine, 2,5-dimethyl, 2,5-dimethyl-benzylamine, 2,5-dimethylbenzenemethanamine, 2,5-dimethylphenyl methylamine, 2,5-dimethylphenyl methylamine, 2-methylamino-p-xylene, 2, 5-dimethylbenzylamine, 2,5-dimethylbenzylam
(2,5-dimethylphenyl)methanamine

Specifications

93-48-1
100.0
97.5% min. (GC)
C9H13N
MFCD00039780
2,5-dimethylbenzylamine, 2,5-dimethylphenyl methanamine, 1-2,5-dimethylphenyl methanamine, benzenemethanamine, 2,5-dimethyl, 2,5-dimethyl-benzylamine, 2,5-dimethylbenzenemethanamine, 2,5-dimethylphenyl methylamine, 2,5-dimethylphenyl methylamine, 2-methylamino-p-xylene, 2, 5-dimethylbenzylamine, 2,5-dimethylbenzylam
CC1=CC(=C(C=C1)C)CN
135.21
135.21
2, 5-Dimethylbenzylamine, 99%
98.5
Authentic
Glass bottle
1.5350 to 1.538
1g
LUJNPFWZXIGIPS-UHFFFAOYSA-N
(2,5-dimethylphenyl)methanamine
66735
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-250-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only