missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-7,7,8,8-tetracyanoquinodimethane 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 500 mg |
| UN Number | 3439 |
| Formula Weight | 232.25 |
| Percent Purity | ≥98.0% (N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,5-Dimethyl-7,7,8,8-tetracyanoquinodimethane |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |