missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-7,7,8,8-tetracyanoquinodimethane 98.0+%, TCI America™

Catalog No. D2021100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2021100MG 100 mg
D2021500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2021100MG Supplier TCI America Supplier No. D2021100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1487-82-7
Molecular Formula C14H8N4
Molecular Weight (g/mol) 232.25
MDL Number MFCD00130123
InChI Key DFJXWQJAMNCPII-UHFFFAOYSA-N
PubChem CID 616047
IUPAC Name 2-[4-(dicyanomethylidene)-2,5-dimethylcyclohexa-2,5-dien-1-ylidene]propanedinitrile
SMILES CC1=CC(=C(C#N)C#N)C(C)=CC1=C(C#N)C#N

Specifications

Quantity 100 mg
UN Number 3439
Formula Weight 232.25
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethyl-7,7,8,8-tetracyanoquinodimethane
TSCA No
Recommended Storage Refrigerator