missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-4-hydroxy-3(2H)-furanone 98.0+%, TCI America™
$76.43 - $199.46
Chemical Identifiers
| CAS | 3658-77-3 |
|---|---|
| Molecular Formula | C6H8O3 |
| Molecular Weight (g/mol) | 128.127 |
| MDL Number | MFCD00010706 |
| InChI Key | INAXVXBDKKUCGI-UHFFFAOYSA-N |
| Synonym | Furaneol, 4-Hydroxy-2,5-dimethyl-3(2H)-furanone |
| PubChem CID | 19309 |
| ChEBI | CHEBI:76247 |
| IUPAC Name | 4-hydroxy-2,5-dimethylfuran-3-one |
| SMILES | CC1C(=O)C(=C(O1)C)O |
Chemical Identifiers
| 3658-77-3 | |
| 128.127 | |
| INAXVXBDKKUCGI-UHFFFAOYSA-N | |
| 19309 | |
| 4-hydroxy-2,5-dimethylfuran-3-one |
| C6H8O3 | |
| MFCD00010706 | |
| Furaneol, 4-Hydroxy-2,5-dimethyl-3(2H)-furanone | |
| CHEBI:76247 | |
| CC1C(=O)C(=C(O1)C)O |
Specifications
| 3658-77-3 | |
| White-Yellow | |
| MFCD00010706 | |
| Furaneol, 4-Hydroxy-2,5-dimethyl-3(2H)-furanone | |
| CC1C(=O)C(=C(O1)C)O | |
| 128.127 | |
| CHEBI:76247 | |
| ≥98.0% (HPLC) | |
| 2,5-Dimethyl-4-hydroxy-3(2H)-furanone |
| 81°C | |
| C6H8O3 | |
| 5 g | |
| INAXVXBDKKUCGI-UHFFFAOYSA-N | |
| 4-hydroxy-2,5-dimethylfuran-3-one | |
| 19309 | |
| 128.13 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-5509
RTECSNumber : LU3990000
TSCA : Yes
Recommended Storage : Refrigerator