missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-4-hydroxy-3(2H)-furanone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 81°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 128.13 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,5-Dimethyl-4-hydroxy-3(2H)-furanone |
Safety and Handling
| EINECSNumber | (5)-5509 |
| RTECSNumber | LU3990000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |