missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2-hexanol, 98%

CAS: 3730-60-7 | C8H18O | 130.23 g/mol

Supplier:  Thermo Scientific Chemicals B2035214

 View more versions of this product

Catalog No. AAB2035214


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,5-Dimethyl-2-hexanol
155°C to 156°C
C8H18O
MFCD00021814
UN1987
2,5-dimethyl-2-hexanol, 2-hexanol, 2,5-dimethyl-, s, 2-hexanol, 2,5-dimethyl, 2-hexanol, 2,5-dimethyl-, s-+, acmc-209is7, 2-hexanol,2,5-dimethyl, 2-hexanol,5-dimethyl-, s, 2-hexanol,5-dimethyl-, s-+
CC(C)CCC(C)(C)O
130.23
130.23
3730-60-7
52°C (125°F)
1.422
25 g
1732761
JPUIYNHIEXIFMV-UHFFFAOYSA-N
2,5-dimethylhexan-2-ol
19506
98%

Chemical Identifiers

3730-60-7
130.23
JPUIYNHIEXIFMV-UHFFFAOYSA-N
19506
CC(C)CCC(C)(C)O
C8H18O
MFCD00021814
2,5-dimethyl-2-hexanol, 2-hexanol, 2,5-dimethyl-, s, 2-hexanol, 2,5-dimethyl, 2-hexanol, 2,5-dimethyl-, s-+, acmc-209is7, 2-hexanol,2,5-dimethyl, 2-hexanol,5-dimethyl-, s, 2-hexanol,5-dimethyl-, s-+
2,5-dimethylhexan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 223-085-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only