missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2-hexanol, 98%

Catalog No. AAB2035206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2035206 5 g
AAB2035214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2035206 Supplier Thermo Scientific Chemicals Supplier No. B2035206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3730-60-7 | C8H18O | 130.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3730-60-7
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00021814
InChI Key JPUIYNHIEXIFMV-UHFFFAOYSA-N
Synonym 2,5-dimethyl-2-hexanol, 2-hexanol, 2,5-dimethyl-, s, 2-hexanol, 2,5-dimethyl, 2-hexanol, 2,5-dimethyl-, s-+, acmc-209is7, 2-hexanol,2,5-dimethyl, 2-hexanol,5-dimethyl-, s, 2-hexanol,5-dimethyl-, s-+
PubChem CID 19506
IUPAC Name 2,5-dimethylhexan-2-ol
SMILES CC(C)CCC(C)(C)O

Specifications

Boiling Point 155°C to 156°C
Flash Point 52°C (125°F)
Refractive Index 1.422
Quantity 5 g
UN Number UN1987
Beilstein 1732761
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 2,5-Dimethyl-2-hexanol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 223-085-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only