missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2,5-hexanediol 99.0+%, TCI America™

Catalog No. p-7126623
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D116725G 25 g
D1167500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D116725G Supplier TCI America Supplier No. D116725G
Only null left

Chemical Identifiers

CAS 110-03-2
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00004473
InChI Key ZWNMRZQYWRLGMM-UHFFFAOYSA-N
Synonym 2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830
PubChem CID 8031
IUPAC Name 2,5-dimethylhexane-2,5-diol
SMILES CC(C)(O)CCC(C)(C)O

Specifications

Melting Point 89°C
Color White
Boiling Point 127°C
Quantity 25 g
Formula Weight 146.23
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethyl-2,5-hexanediol
EINECSNumber (2)-0240
TSCA Yes