missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2,5-hexanediol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 162500050

Catalog No. AC162500050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 5-Dimethyl-2, 5-hexanediol
110-03-2
98.5
214°C to 215°C
Conforms
Glass bottle
HOC(CH3)2CH2CH2C(CH3)2OH
MFCD00004473
2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830
ZWNMRZQYWRLGMM-UHFFFAOYSA-N
2,5-dimethylhexane-2,5-diol
8031
97%
97%
GC≥96.0%
100.0
126°C
96% min. (GC)
C8H18O2
5g
01, 492
Solubility in water: soluble. Other solubilities: soluble in acetone,ethylene glycol,insoluble in benzene,ccl4,petroleuw ether
CC(C)(O)CCC(C)(C)O
146.23
146.23
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS Signal Word: Danger

EINECSNumber : 203-731-5

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions